Dipropyleneglycol monomethyl ether (mixture of isomers) mixture of isomers ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:34590-94-8
:
Formula:C₇H₁₆O₃
MDL Number:MFCD00059604
:148.2001 g/mol
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Assay (GC, area%) (isomers):≥ 99,0 %
- Identification (IR):Passes test
- Density (20/4):0,945 - 0,957
- Water :≤ 0,15 %
Specification Test Results
Related Information
Frequently Bought Together