Anti-DR3 (TRAMP) Mouse Monoclonal Antibody [clone:JD3](TotalSeq™-B1057)
Catalog # BLEG307109
Supplier: BIOLEGEND INC
Specifications
- Antigen symbol:DR3 (TRAMP)
- Clonality:Monoclonal
- Clone:JD3
- Conjugation:TotalSeq™-B1057
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen synonyms:TNFRSF25, TRAMP, Apo-3, WSL-1, LARD, DDR3, TR3, TNFRSF12
- Storage temperature:2…8 °C
- Concentration:0.5 mg/ml
- Immunogen:human DR3-Ig fusion protein
- Purification:The antibody was purified by chromatography and conjugated with TotalSeq™-B oligomer under optimal conditions.
- Pk:10 µG