Mouse recombinant TRANCE (RANKL, TNFSF11) (from E.coli)
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20…–70 °C
- Carrier-free:Y
- Protein synonyms:member 11 (TNFSF11)|TNF-related activation-induced cytokine (Trance)|Osteoprotegerin ligand (OPGL)|Tumor necrosis factor ligand superfamily|Receptor activator of NF-kappa-B ligand (RANKL)|Osteoclast differentiation factor (ODF)|CD254.
- Protein/peptide name:TRANCE (RANKL, TNFSF11)
- Concentration:0,2 mg/ml
- Formulation:Sterile-filtered with 0,2 µm filter, solution comprised of 20 mM sodium phosphate, pH 6,0, 100 mM NaCl, and 2 mM EDTA
- Tested applications:BA