Human recombinant NRG1-beta1/HRG1-beta1 ECD domain (from E.coli)
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20…–70 °C
- Carrier-free:Y
- Protein synonyms:Heregulin-1 beta1 (HRG1-beta1).|Neuregulin-1 beta1 (NRG1-beta1)
- Protein/peptide name:NRG1-beta1/HRG1-beta1 ECD domain
- Concentration:0,2 mg/ml
- Formulation:0,22 µm filtered protein solution is in 1X PBS
- Tested applications:BA