Human Recombinant Fc gamma RIIIA/CD16a
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:293E cells
- Species:Human
- Storage conditions:–20...–70 °C
- Carrier-free:Y
- Protein synonyms:Fc-gamma RIIIa|FcRIIIa|IgG Fc receptor III-2|Fc-gamma RIII|FcR-10|FCGR3A|CD16a antigen|Fc-gamma RIII-alpha|Low affinity immunoglobulin gamma Fc region receptor III-A|FCGR3|FcRIII|CD16a|IGFR3
- Protein/peptide name:Fc gamma RIIIA/CD16a
- Formulation:0,22 µm filtered protein solution is in PBS, pH 7,2
- Tested applications:BA
- Concentration:0,2 mg/ml