Rat recombinant erythropoietin (EPO) from CHO cells
Supplier: BIOLEGEND INC
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Rat
- Storage conditions:–20…–70 °C
- Carrier-free:Y
- Protein synonyms:EP|Hematopoietin|MVCD2
- Protein/peptide name:Erythropoietin (EPO)
- Concentration:0,2 mg/ml
- Formulation:0,22 µm filtered protein solution is in 1X PBS, pH 7,2
- Tested applications:BA
Specifications
Frequently Bought Together