Human recombinant CEACAM-8/CD66b
Marketsource
Supplier: BIOLEGEND INC
Über diesen Artikel
4 mögliche Produktvariationen vorhanden
Product Details & Documents
Spezifikationen für diese Produktfamilie
Spezifikationen:
Spezifikationen:
- Catalog No:
- BLEG778102
- BLEG778104
- BLEG778106
- BLEG778108
- Protein/peptide name:
- CEACAM-8/CD66b
- CEACAM-8/CD66b
- CEACAM-8/CD66b
- CEACAM-8/CD66b
- Protein synonyms:
- CEACAM-8|Carcinoembryonic antigen CGM6|CD67 antigen|CEACAM8|CD66b|Carcinoembryonic antigen-related cell adhesion molecule 8|Nonspecific cross-reacting antigen NCA-95
- CEACAM-8|Carcinoembryonic antigen CGM6|CD67 antigen|CEACAM8|CD66b|Carcinoembryonic antigen-related cell adhesion molecule 8|Nonspecific cross-reacting antigen NCA-95
- CEACAM-8|Carcinoembryonic antigen CGM6|CD67 antigen|CEACAM8|CD66b|Carcinoembryonic antigen-related cell adhesion molecule 8|Nonspecific cross-reacting antigen NCA-95
- CEACAM-8|Carcinoembryonic antigen CGM6|CD67 antigen|CEACAM8|CD66b|Carcinoembryonic antigen-related cell adhesion molecule 8|Nonspecific cross-reacting antigen NCA-95
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Human
- Human
- Source:
- 293E cells
- 293E cells
- 293E cells
- 293E cells
- Carrier-free:
- Y
- Y
- Y
- Y
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- 0,2 mg/ml
- 0,2 mg/ml
- Formulation:
- 0,22 µm filtered protein solution is in PBS, pH 7,2
- 0,22 µm filtered protein solution is in PBS, pH 7,2
- 0,22 µm filtered protein solution is in PBS, pH 7,2
- 0,22 µm filtered protein solution is in PBS, pH 7,2
- Storage conditions:
- –20...–70 °C
- –20...–70 °C
- –20...–70 °C
- –20...–70 °C
- Tested applications:
- BA
- BA
- BA
- BA
Spezifikationen:
Product Family Options
Produktinformationen
- PkAvailabilityPrice
- Spezifikationen:Marketsource
More
Specifications
Cat. No.BLEG778102Protein/peptide nameCEACAM-8/CD66bProtein synonymsCEACAM-8|Carcinoembryonic antigen CGM6|CD67 antigen|CEACAM8|CD66b|Carcinoembryonic antigen-related cell adhesion molecule 8|Nonspecific cross-reacting antigen NCA-95Protein/peptide typeRecombinantSpeciesHumanSource293E cellsCarrier-freeYConcentration0,2 mg/mlFormulation0,22 µm filtered protein solution is in PBS, pH 7,2Storage conditions–20...–70 °CTested applicationsBAMarketsource - Spezifikationen:Marketsource
More
Specifications
Cat. No.BLEG778104Protein/peptide nameCEACAM-8/CD66bProtein synonymsCEACAM-8|Carcinoembryonic antigen CGM6|CD67 antigen|CEACAM8|CD66b|Carcinoembryonic antigen-related cell adhesion molecule 8|Nonspecific cross-reacting antigen NCA-95Protein/peptide typeRecombinantSpeciesHumanSource293E cellsCarrier-freeYConcentration0,2 mg/mlFormulation0,22 µm filtered protein solution is in PBS, pH 7,2Storage conditions–20...–70 °CTested applicationsBAMarketsource - Spezifikationen:Marketsource
More
Specifications
Cat. No.BLEG778106Protein/peptide nameCEACAM-8/CD66bProtein synonymsCEACAM-8|Carcinoembryonic antigen CGM6|CD67 antigen|CEACAM8|CD66b|Carcinoembryonic antigen-related cell adhesion molecule 8|Nonspecific cross-reacting antigen NCA-95Protein/peptide typeRecombinantSpeciesHumanSource293E cellsCarrier-freeYConcentration0,2 mg/mlFormulation0,22 µm filtered protein solution is in PBS, pH 7,2Storage conditions–20...–70 °CTested applicationsBAMarketsource - Spezifikationen:Marketsource
More
Specifications
Cat. No.BLEG778108Protein/peptide nameCEACAM-8/CD66bProtein synonymsCEACAM-8|Carcinoembryonic antigen CGM6|CD67 antigen|CEACAM8|CD66b|Carcinoembryonic antigen-related cell adhesion molecule 8|Nonspecific cross-reacting antigen NCA-95Protein/peptide typeRecombinantSpeciesHumanSource293E cellsCarrier-freeYConcentration0,2 mg/mlFormulation0,22 µm filtered protein solution is in PBS, pH 7,2Storage conditions–20...–70 °CTested applicationsBAMarketsource



