Rat recombinant CNTF
Marketsource
- Catalog No:
- BLEG776202
- BLEG776204
- BLEG776206
- BLEG776208
- Protein/peptide name:
- CNTF
- CNTF
- CNTF
- CNTF
- Protein synonyms:
- Ciliary neurotrophic factor
- Ciliary neurotrophic factor
- Ciliary neurotrophic factor
- Ciliary neurotrophic factor
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Species:
- Rat
- Rat
- Rat
- Rat
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- Carrier-free:
- Y
- Y
- Y
- Y
- Concentration:
- 0,2 mg/ml
- 0,2 mg/ml
- 0,2 mg/ml
- 0,2 mg/ml
- Formulation:
- 0,22 µm filtered protein solution is in 20 mM Tris, 150 mM NaCl, 1 mM EDTA, 2 mM CHAPS, 2 mM TCEP, pH 7,5
- 0,22 µm filtered protein solution is in 20 mM Tris, 150 mM NaCl, 1 mM EDTA, 2 mM CHAPS, 2 mM TCEP, pH 7,5
- 0,22 µm filtered protein solution is in 20 mM Tris, 150 mM NaCl, 1 mM EDTA, 2 mM CHAPS, 2 mM TCEP, pH 7,5
- 0,22 µm filtered protein solution is in 20 mM Tris, 150 mM NaCl, 1 mM EDTA, 2 mM CHAPS, 2 mM TCEP, pH 7,5
- Storage conditions:
- –20...–70 °C
- –20...–70 °C
- –20...–70 °C
- –20...–70 °C
- Tested applications:
- BA
- BA
- BA
- BA
Product Family Options
- Pk
- Marketsource
Specifications
Cat. No.BLEG776202Protein/peptide nameCNTFProtein synonymsCiliary neurotrophic factorProtein/peptide typeRecombinantSpeciesRatSourceE. coliCarrier-freeYConcentration0,2 mg/mlFormulation0,22 µm filtered protein solution is in 20 mM Tris, 150 mM NaCl, 1 mM EDTA, 2 mM CHAPS, 2 mM TCEP, pH 7,5Storage conditions–20...–70 °CTested applicationsBAMarketsource - Marketsource
Specifications
Cat. No.BLEG776204Protein/peptide nameCNTFProtein synonymsCiliary neurotrophic factorProtein/peptide typeRecombinantSpeciesRatSourceE. coliCarrier-freeYConcentration0,2 mg/mlFormulation0,22 µm filtered protein solution is in 20 mM Tris, 150 mM NaCl, 1 mM EDTA, 2 mM CHAPS, 2 mM TCEP, pH 7,5Storage conditions–20...–70 °CTested applicationsBAMarketsource - Marketsource
Specifications
Cat. No.BLEG776206Protein/peptide nameCNTFProtein synonymsCiliary neurotrophic factorProtein/peptide typeRecombinantSpeciesRatSourceE. coliCarrier-freeYConcentration0,2 mg/mlFormulation0,22 µm filtered protein solution is in 20 mM Tris, 150 mM NaCl, 1 mM EDTA, 2 mM CHAPS, 2 mM TCEP, pH 7,5Storage conditions–20...–70 °CTested applicationsBAMarketsource - Marketsource
Specifications
Cat. No.BLEG776208Protein/peptide nameCNTFProtein synonymsCiliary neurotrophic factorProtein/peptide typeRecombinantSpeciesRatSourceE. coliCarrier-freeYConcentration0,2 mg/mlFormulation0,22 µm filtered protein solution is in 20 mM Tris, 150 mM NaCl, 1 mM EDTA, 2 mM CHAPS, 2 mM TCEP, pH 7,5Storage conditions–20...–70 °CTested applicationsBAMarketsource



