Order Entry
Germany
ContactUsLinkComponent
Orthophosphoric acid ≥98%, AnalaR NORMAPUR®
Orthophosphoric acid ≥98%, AnalaR NORMAPUR®
Catalog # 89035.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Orthophosphoric acid ≥98%, AnalaR NORMAPUR®
Catalog # 89035.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (GC):
    min. 99.9 %
  • Non-volatile matter:
    max. 0.0005 %
  • Water (Karl Fischer):
    max. 0.05 %
  • Suitability for residue analysis:
    max. 5 ng/l
  • Vapor Pressure:
    72.8 mmHg (20 °C)
  • Explosion Limit:
    0.16
  • Assay:
    min 98.0%
  • Identification:
    Passes test
  • Substances reducing KMnO₄ (as O):
    max 20 ppm
  • Volatile acids (as CH₃COOH):
    max 20 ppm
  • Cl (Chloride):
    max 1 ppm
  • F (Fluoride):
    max 0.2 ppm
  • NO₃ (Nitrate):
    max 2 ppm
  • PO₃ + PO₂ (as H₃PO₃):
    max 0.05%
  • SO₄ (Sulphate):
    max 30 ppm
  • As (Arsenic):
    max 1 ppm
  • Ca (Calcium):
    max 10 ppm
  • Cd (Cadmium):
    max 1 ppm
  • Co (Cobalt):
    max 1 ppm
  • Cr (Chromium):
    max 1 ppm
  • Cu (Copper):
    max 1 ppm
  • Fe (Iron):
    max 10 ppm
  • K (Potassium):
    max 5 ppm
  • Mg (Magnesium):
    max 1 ppm
  • Mn (Manganese):
    max 1 ppm
  • Na (Sodium):
    max 5 ppm
  • Ni (Nickel):
    max 1 ppm
  • Pb (Lead):
    max 1 ppm
  • Sb (Antimony):
    max 1 ppm
  • Zn (Zinc):
    max 1 ppm

Specification Test Results

About this item

 :
 :98 g/mol
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340