Orders LinkContactUsLinkComponent
General purpose gloves, Perfect Poly® Skin
General purpose gloves, Perfect Poly® Skin
Supplier:   HONEYWELL SAFETY
General purpose gloves, Perfect Poly® Skin
Supplier:  HONEYWELL SAFETY
Certificates
General purpose gloves, Perfect Poly® Skin
4 Options Available Below
Product Details & Documents
Polyamide with its polyurethane coating provide excellent abrasion and tear resistance. Non pilling, the polyamide is suitable for precision work. The ventilated back and ultra light coating optimise the glove's breathability.
  • Offers maximum comfort and added sturdiness
  • The knitting technique helps to obtain a glove that conforms snugly to the hand as well as outstanding dexterity
  • The ultra tight weave gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • Certified as 'silicone-free' to ensure their use in painting
  • Its blue colour and grey coating make the glove more soilproof, thus ensuring a longer life
Ideal for the handling of fine products in a dry or slightly greasy or soiled environment.
Gauge: 15
Colour: blue/grey
EN 388 (2121), Cat. III
Certifications: EN 388:2016 (2110X); Cat. II
Specifications for Product Family
Specifications
  • Catalog No:
  • 112-1649
  • 112-1650
  • 112-1651
  • 112-1648
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Finish:
  • Coating on palm and fingertips
  • Coating on palm and fingertips
  • Coating on palm and fingertips
  • Coating on palm and fingertips
  • Gauge:
  • 18
  • 18
  • 18
  • 18
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (2110X)
  • EN 388:2016 (2110X)
  • EN 388:2016 (2110X)
  • EN 388:2016 (2110X)
Specifications
Product Family Options
Product Information
  • Glove size
    Colour
    Length
    Availability
    Price
  • 7
    Blue/grey
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 27,90 €
    27,90 €
    1 * 10 PAAR
       27,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-1649
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Finish
    Coating on palm and fingertips
    Gauge
    18
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (2110X)
  • 8
    Blue/grey
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 27,90 €
    27,90 €
    1 * 10 PAAR
       27,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-1650
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Finish
    Coating on palm and fingertips
    Gauge
    18
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (2110X)
  • 9
    Blue/grey
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 27,90 €
    27,90 €
    1 * 10 PAAR
       27,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-1651
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Finish
    Coating on palm and fingertips
    Gauge
    18
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (2110X)
  • 10
    Blue/grey
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 PAAR
    1 * 10 PAAR - 27,90 €
    27,90 €
    1 * 10 PAAR
       27,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-1648
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Liner material
    Polyamide
    Finish
    Coating on palm and fingertips
    Gauge
    18
    Cuff style
    Knit wrist
    EN norm
    EN 388:2016 (2110X)