Orders LinkContactUsLinkComponent
Headspace sampler supplies
Supplier:   Agilent
Headspace sampler supplies
Supplier:  Agilent
Certificates
Headspace sampler supplies
Headspace sampler supplies
18 Options Available Below
Product Details & Documents
Find the headspace supplies you need for your Agilent 7697 or G1888 headspace sampler.
  • Built-in pneumatics for superior control and easier setup
  • Proven valve and loop sampling technology
  • Fully-automatic sample vial leak checking and available bar code reader help ensure greater confidence in results method compatibility
  • Instrument control software that is fully integrated in data systems
  • Resource conserving programmable instrument scheduler
Headspace sampling allows introduction of volatile compounds to the GC or GC/MS from virtually any sample matrix while leaving unwanted components in a disposable sample vial. This results in less maintenance and longer lifetimes for the GC consumables in your lab. With up to 111 sample vial positions and removable vial racks, the 7697A's inert sample pathway supports nearly continuous operation to satisfy even the busiest laboratory. Alternatively, it's legacy G1888 headspace sampler also features an inert sample pathway for superior chemical performance without analyte degradation or loss.
Product Family Options
Product Information
  • Availability
    Price
  • HEWLG4556-90500
    7697A Tray labels
    7697A Tray labels
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • HEWLG4556-60019
    7697A Tray rack
    7697A Tray rack
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 106,00 €
    106,00 €
    1 * 1 ST
       106,00 €
  • HEWL0100-2595
    Ferrule polyamide graphite, ¹/₃₂"
    Ferrule polyamide graphite, ¹/₃₂"
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 86,00 €
    86,00 €
    1 * 5 ST
       86,00 €
  • HEWLG4556-67010
    Leak test kit (ferrule - no hole, plug ¹/₈" tube fitting, nylon, headspace leak test vial, plug ¹/₁₆" ZDV stainless steel male, 11 mm low bleed septa)
    Leak test kit (ferrule - no hole, plug ¹/₈" tube fitting, nylon, headspace leak test vial, plug ¹/₁₆" ZDV stainless steel male, 11 mm low bleed septa)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 178,00 €
    178,00 €
    1 * 1 ST
       178,00 €
  • HEWL4177-0607
    ProSteel sleeve
    ProSteel sleeve
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,00 €
    43,00 €
    1 * 1 ST
       43,00 €
  • HEWLG4556-80101
    Sample loop, headspace 0,025 ml, SN 2000
    Sample loop, headspace 0,025 ml, SN 2000
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 634,00 €
    634,00 €
    1 * 1 ST
       634,00 €
  • HEWLG4556-80102
    Sample loop, headspace 0,05 ml, SN 2000
    Sample loop, headspace 0,05 ml, SN 2000
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 634,00 €
    634,00 €
    1 * 1 ST
       634,00 €
  • HEWLG4556-80103
    Sample loop, headspace 0,10 ml, SN2000
    Sample loop, headspace 0,10 ml, SN2000
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 634,00 €
    634,00 €
    1 * 1 ST
       634,00 €
  • HEWLG4556-80105
    Sample loop, headspace 0,50 ml, SN2000
    Sample loop, headspace 0,50 ml, SN2000
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 634,00 €
    634,00 €
    1 * 1 ST
       634,00 €
  • HEWLG4556-80106
    Sample loop, headspace 1,0 ml, SN2000
    Sample loop, headspace 1,0 ml, SN2000
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 634,00 €
    634,00 €
    1 * 1 ST
       634,00 €
  • HEWLG4556-80108
    Sample loop, headspace 3,00 ml, SN2000
    Sample loop, headspace 3,00 ml, SN2000
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 634,00 €
    634,00 €
    1 * 1 ST
       634,00 €
  • HEWLG4556-80109
    Sample loop, headspace 5,00 ml, SN2000
    Sample loop, headspace 5,00 ml, SN2000
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 634,00 €
    634,00 €
    1 * 1 ST
       634,00 €
  • HEWLG4556-80126
    Sample loop, HS, certified, 1,00 ml, SN2000
    Sample loop, HS, certified, 1,00 ml, SN2000
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 765,00 €
    765,00 €
    1 * 1 ST
       765,00 €
  • HEWLG4556-80128
    Sample loop, HS, certified, 3,00 ml, SN2000
    Sample loop, HS, certified, 3,00 ml, SN2000
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 765,00 €
    765,00 €
    1 * 1 ST
       765,00 €
  • HEWLG4556-80107
    Sample loop, inert, 2 ml, for Agilent 7697A headspace sampler
    Sample loop, inert, 2 ml, for Agilent 7697A headspace sampler
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 671,00 €
    671,00 €
    1 * 1 ST
       671,00 €
  • HEWLG3452-60835
    Septum retainer nut for transfer line
    Septum retainer nut for transfer line
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 112,00 €
    112,00 €
    1 * 1 ST
       112,00 €
  • HEWLG3520-20210
    Transfer line nut fitting for G3520A transfer line interface accessory
    Transfer line nut fitting for G3520A transfer line interface accessory
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 90,00 €
    90,00 €
    1 * 1 ST
       90,00 €
  • HEWLRDT-1020
    Trap, SplitVent and 3 cartridges
    Trap, SplitVent and 3 cartridges
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 215,00 €
    215,00 €
    1 * 3 ST
       215,00 €