Order Entry
Germany
ContactUsLinkComponent
VWR®, Real-Time PCR kit for Salmonella spp. in food samples
VWR®, Real-Time PCR kit for Salmonella spp. in food samples
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Real-Time PCR kit for Salmonella spp. in food samples
Supplier:  VWR Chemicals
Certificates

Specifications

  • Application:
    qPCR probe
  • Storage conditions:
    stored in a freezer at -20ºC

Specifications

DNA extraction kit for food samples

Product Information