GC columns, J&W HP-35
Lieferant: Agilent
Zertifikate
Über diesen Artikel
HP-35 is a mid-polarity column, inert to active solutes and ideal for conformational analyses.
- (35%-Phenyl)-methylpolysiloxane
- Mid polarity-slightly less polar than DB-35
- Inert to active solutes
- Ideal for confirmational analyses
- Bonded and cross-linked
This phase has been specified by EPA method 8081 and U.S.P G-42. The intermediate polarity of HP-35 makes it a good alternative for the analysis of pesticides, herbicides, pharmaceuticals, and amines.
Frequently Bought Together