ContactUsLinkComponent
Glass beakers, low form, Super Duty, DURAN®
Glass beakers, low form, Super Duty, DURAN®
Catalog # 214-0204
Supplier:  DWK Life Sciences
Supplier #: 211073605
undefined
Glass beakers, low form, Super Duty, DURAN®
Catalog # 214-0204
Supplier:  DWK Life Sciences
Supplier #: 211073605
Log in for availability
Select an Option
70 mm
Select an Option
250 ml
Select an Option
95 mm
Select an Option
02
Select an Option
1
About this item
DURAN®, borosilicate glass 3.3, clear, with spout.
  • Higher mechanical stability compared with standard glass beakers due to reinforced sturdy rim
  • Even distribution of wall thickness
  • Very good chemical resistance, good thermal resistance
With easily legible graduation and labelling area. It is recommended that you heat Super Duty glass beakers slowly and evenly to prevent thermal stresses in the glass. For work at very high temperatures, or with fast changes in temperature, standard DURAN® beakers should be used, as these offer excellent resistance to temperature changes. However, mechanical stability is limited with these products in comparison with the Super Duty beakers.
With printed Retrace Code (batch identification).