Capillary tubing
Supplier: Agilent
Certificates
About this item
A wide range of capillary tubing, rigid capillary is available.
- Flexible and easy to cut to desired lengths
- Color coded for easy tracking
- Accepts both stainless steel fittings
- Ext. Ø: 1,58 mm