Vacin and Went modified orchid basal salt mixture
Contains an equivalent iron molar concentration of ferrous sulfate in place of ferric tartrate.
- Plant tissue culture tested
Contains an equivalent iron molar concentration of ferrous sulfate in place of ferric tartrate.