Nitsch and Nitsch basal medium with vitamins
Supplier: Apollo Scientific
Certificates
About this item
Frequently Bought Together
n-Hexane ≥95%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
(±)-2-Methyltetrahydrofuran ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pyridine, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cobalt(II) chloride hexahydrate, redish purple crystals
Supplier: MP Biomedicals