GC columns, J&W DX-1 and DX-4
Supplier: Agilent
Certificates
About this item
DX-Series of GC columns, the polyethylene glycol (PEG) DX-1 and DX-4 columns, deliver excellent performance over a wide temperature range.
- DX-1: 90% dimethylpolysiloxane 10% polyethylene glycol
- DX-4: 15% dimethylpolysiloxane 85% polyethylene glycol
- Each Agilent J&W GC column is tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency to provide you utmost confidence in qualitative and quantitative results
- For details about the QC test conditions including the test probe mixture and the resulting chromatogram, please refer to the Agilent J&W GC Column Performance Summary shipped with each GC column. Because DX series GC columns are not bonded and cross-linked, we do not recommend solvent rinsing
The DX series columns operate within temperature limits of 50 to 250/270 °C.
Caution: DX series columns are not bonded and cross-linked, and so we do not recommend solvent rinsing.