Taq extender PCR additive
Catalog # AGLS600148
Supplier: AGILENT
CAS Number:
Specifications
- Pk:1
- Description:Taq extender PCR additive, 1000 U
- Details:Taq extender is used to improve reliability of and yield of conventional PCR. This additive increases efficiency at which Taq performs and also improves PCR amplification of difficult templates and increases reliability and yield of almost any PCR target up to 10 kb in length.
Specifications
About this item
Improves reliability and yield of conventional Taq-based PCR amplifications.
- Improves length, yield, and reliability of PCR
- Enhances efficiency of template extension by Taq DNA polymerase
- Improves amplification of difficult targets
Taq Extender PCR additive improves the reliability and yield of conventional Taq-based PCR amplifications. Taq Extender additive increases the efficiency of Taq DNA polymerase extension reactions during each cycle of PCR by reducing mismatch pausing, resulting in a greater percentage of completed extension reactions.
Caution: For research use only. Not for use in diagnostic procedures.