Ampicillin
200×25 mg, 1
Catalog # STRA300021
Marketsource
Supplier: STRATAGENE
Jump to:
Product Details & Documents
Amp Tabs Ampicillin Tablets are used to select for ampicillin-resistant bacteria and turbo Amp Antibiotic is used to reduce satellite colonies when selecting for ampicillin-resistant bacteria.
: For Research Use Only. Not for Use in Diagnostic Procedures.
- CAS Number:69-53-4
- Formula:C₁₆H₁₉N₃O₄S
- MDL Number:MFCD00005175
- Molecular Weight:349.4 g/mol
Specifications
Specifications
- Pk:1
- Pk Info:200×25 mg



