Tubing
Über diesen Artikel
As part of the MINIPULS® 3 and Evolution lines of peristaltic pumps, Gilson offers a variety of tubing of different material, inner diameter sizes, and lengths.
Silicone (Versilic®) tubing is biologically neutral and does not inhibit cell culture while meeting European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
ISO-VERSINIC® tubing shows excellent resistance to strong acids, oxidising agents, and aromatic and chlorinated solvents.
PharMed® tubing meets European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
PVC (Tygon®) tubing handles virtually all inorganic chemicals found in the lab and can be used in corrosive environments.
55 mögliche Produktvariationen vorhanden
Product Details & Documents
As part of the MINIPULS® 3 and Evolution lines of peristaltic pumps, Gilson offers a variety of tubing of different material, inner diameter sizes, and lengths.
Silicone (Versilic®) tubing is biologically neutral and does not inhibit cell culture while meeting European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
ISO-VERSINIC® tubing shows excellent resistance to strong acids, oxidising agents, and aromatic and chlorinated solvents.
PharMed® tubing meets European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
PVC (Tygon®) tubing handles virtually all inorganic chemicals found in the lab and can be used in corrosive environments.
Product Family Options
ISO-VERSINIC®
Produktinformationen
- Tubing int.ØTubing ext.ØLengthColour codeAvailabilityPrice
- GILSF1817741Reference tubing
- GILSF1817743Reference tubing
- GILSF1817745Reference tubing
- GILSF117749Reference tubing
- GILSF117740Extension tubing
- GILSF117742Extension tubing
- GILSF117744Extension tubing
- GILSF117748Extension tubing
- GILSF117750Extension tubing
PVC (Tygon®)
Produktinformationen
- Tubing int.ØTubing ext.ØLengthColour codeAvailabilityPrice
- GILSF117933Reference tubing
- GILSF117934Reference tubing
- GILSF117935Reference tubing
- GILSF117936Reference tubing
- GILSF117938Reference tubing
- GILSF117939Reference tubing
- GILSF117940Reference tubing
- GILSF117941Reference tubing
- GILSF117942Reference tubing
- GILSF117943Reference tubing
- GILSF117945Reference tubing (purple/purple colour code)Reference tubing (purple/purple colour code)2,06 mm (0,081")0,4 mPurpleAnmelden für die Verfügbarkeit1 * 12 ST71,00 €
- GILSF117946Reference tubing (Purple/black colour code)Reference tubing (Purple/black colour code)2,29 mm (0,090")0,4 mPurpleAnmelden für die Verfügbarkeit1 * 12 ST75,00 €
- GILSF117948Reference tubing
- GILSF117949Reference tubing
- GILSF117952Extension tubing
- GILSF117953Extension tubing
- GILSF117954Extension tubing
- GILSF117956Extension tubing
- GILSF117958Extension tubing
- GILSF117960Extension tubing
- GILSF117962Extension tubing
- GILSF117963Extension tubing
- GILSF117965Extension tubing
- GILSF117966Extension tubing
- GILSF117968Extension tubing
- GILSF117969Extension tubing
- GILSF117970Extension tubing
- GILSF117980Extension tubing
- GILSF117981Extension tubing
- GILSF117982Extension tubing
Silicone (Versilic®)
Produktinformationen
- Tubing int.ØTubing ext.ØLengthColour codeAvailabilityPrice
- GILSF1825111Reference tubing
- GILSF1825112Reference tubing
- GILSF1825113Reference tubing (purple/purple colour code)Reference tubing (purple/purple colour code)2,06 mm (0,081")0,4 mPurpleAnmelden für die Verfügbarkeit1 * 4 ST89,00 €
- GILSF1825114Reference tubing
- GILSF1825131Extension tubing
- GILSF1825132Extension tubing
- GILSF1825133Extension tubing
- GILSF117975Extension tubing
- GILSF117976Extension tubing
- GILSF117977Extension tubing
- GILSF117978Extension tubing
- GILSF117979Extension tubing
PharMed®
Produktinformationen
- Tubing int.ØTubing ext.ØLengthColour codeAvailabilityPrice
- GILSF1825101Reference tubing
- GILSF1825102Reference tubing
- GILSF1825103Reference tubing (purple/purple colour code)Reference tubing (purple/purple colour code)2,06 mm (0,081")0,4 mPurpleAnmelden für die Verfügbarkeit1 * 4 ST106,00 €
- GILSF1825122Extension tubing