Orders LinkContactUsLinkComponent
Lambda ZAP II vector kits
Lambda ZAP II vector kits
LambdaZAP II/EcoR l/Gigapack III gold kit, 1
Catalog # STRA236612
Marketsource
Supplier:   STRATAGENE
Lambda ZAP II vector kits
Catalog # STRA236612
Supplier:  STRATAGENE
Marketsource
CAS Number:  
Lambda ZAP II vector kits
LambdaZAP II/EcoR l/Gigapack III gold kit, 1
CAS Number:  
Price
2 440,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The Lambda ZAP II vectors combine the high efficiency of lambda vector systems with the versatility of plasmid systems.
They enable prokaryotic expression of your gene of interest under control of the lac promoter.
in vivo excision of the Lambda ZAP II vector yields the pBluescript SK- vector, which is a high-copy number pUC-based phagemid.
Specifications
Specifications
  • Pk:
    1
  • Description:
    LambdaZAP II/EcoR l/Gigapack III gold kit
Specifications