Restriction enzymes, Dpn I restriction enzymes
Catalog # AGLS500402
Supplier: AGILENT
CAS Number:
Specifications
- Pk:1
- Description:Dpn I
- Units:200 units
Specifications
About this item
Dpn I is an ideal enzyme for cloning and southern blot analysis.
- High purity enzyme preparation
Delivery: Includes 10X universal reaction buffer.
Caution: For research use only. Not for use in diagnostic procedures.