Trans-reporting systems, PathDetect
About this item
PathDetect trans-reporting systems allow you to determine if the protein or compound of interest directly or indirectly activates one of six transcription factors: c-Jun, Elk1, CREB, c-Fos, ATF2 or CHOP.
- Identify pathway-specific transcription activation
- Easier and faster than blotting or gel-shift assays
- Sensitive, quantitative assays
Easily investigate whethe the protein or compound influences the activation of any of the six transcription factors offered and etermine the direct or indirect involvement of new gene products, growth factors, and drug candidates in specific pathways.
Delivery information: Contains of a reporter plasmid, a trans-activator plasmid and both positive and negative control plasmids.
Caution: For research use only. Not for use in diagnostic procedures.
20 Options Available Below
Product Details & Documents
PathDetect trans-reporting systems allow you to determine if the protein or compound of interest directly or indirectly activates one of six transcription factors: c-Jun, Elk1, CREB, c-Fos, ATF2 or CHOP.
- Identify pathway-specific transcription activation
- Easier and faster than blotting or gel-shift assays
- Sensitive, quantitative assays
Easily investigate whethe the protein or compound influences the activation of any of the six transcription factors offered and etermine the direct or indirect involvement of new gene products, growth factors, and drug candidates in specific pathways.
Delivery information: Contains of a reporter plasmid, a trans-activator plasmid and both positive and negative control plasmids.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Product Information
- SizeDetailsPkAvailabilityPrice
- STRA219026PathDetect pFA-ATF2 plasmidPathDetect pFA-ATF2 plasmid10 μgFusion trans-activator plasmid1Log in for availability1 * 1 ST721,00 €Specifications:MarketsourceMarketsource
- STRA219031PathDetect pFA-cFos plasmidPathDetect pFA-cFos plasmid10 μgFusion trans-activator plasmid1Log in for availability1 * 1 ST721,00 €Specifications:MarketsourceMarketsource
- STRA219040PathDetect pMUT-ELK1 plasmidPathDetect pMUT-ELK1 plasmid10 μgNegative control plasmid for ELK1 reporting system10 µGLog in for availability1 * 10 µG831,00 €Specifications:MarketsourceMarketsource
- STRA219041PathDetect pMUT-cJun plasmidPathDetect pMUT-cJun plasmid10 μgNegative control plasmid for c-Jun reporting system10 µGLog in for availability1 * 10 µG721,00 €Specifications:MarketsourceMarketsource
- STRA219042PathDetect pMUT-CREB plasmidPathDetect pMUT-CREB plasmid10 μgNegative control plasmid for the CREB reporting system10 µGLog in for availability1 * 10 µG831,00 €Specifications:MarketsourceMarketsource
- STRA219053PathDetect pFA-Jun plasmidPathDetect pFA-Jun plasmid10 μgActivator plasmid for c-Jun reporting system1Log in for availability1 * 1 ST721,00 €Specifications:MarketsourceMarketsource
- STRA219054PathDetect pFA-CHOP plasmidPathDetect pFA-CHOP plasmid10 μgFusion transactivator plasmid for CHOP reporting system1Log in for availability1 * 1 ST721,00 €Specifications:MarketsourceMarketsource
- STRA219056PathDetect pFC2-dbd Trans-reporter control plasmidPathDetect pFC2-dbd Trans-reporter control plasmid10 μgNegative control plasmid for all PathDetect trans-systems1Log in for availability1 * 1 ST721,00 €Specifications:MarketsourceMarketsource
- STRA219062PathDetect pFA-Elk1 plasmidPathDetect pFA-Elk1 plasmid10 μgActivator plasmid for Elk1 reporting system10 µGLog in for availability1 * 10 µG721,00 €Specifications:MarketsourceMarketsource
- STRA219065PathDetect pFC-MEK1 plasmidPathDetect pFC-MEK1 plasmid10 μgPositive control plasmid for Elk1 reporting system1Log in for availability1 * 1 ST721,00 €Specifications:MarketsourceMarketsource
- STRA219068PathDetect pFA-CREB plasmidPathDetect pFA-CREB plasmid10 μgActivator plasmid for CREB reporting system1Log in for availability1 * 1 ST721,00 €Specifications:MarketsourceMarketsource
- STRA219086PathDetect pFC-MEK3 plasmidSpecifications:MarketsourceMarketsource
- STRA219036PathDetect pFA-CMV plasmidPathDetect pFA-CMV plasmid20 μgFusion trans-activator cloning vector. Create your own trans reporting vector with the pathway specific activator of your choice20 µGLog in for availability1 * 20 µG885,00 €Specifications:MarketsourceMarketsource
- STRA219001PathDetect pFR-CAT Trans-reporter plasmidPathDetect pFR-CAT Trans-reporter plasmid50 μgChloroamphenicol acetyltransferase reporter plasmid. Indicates activation of upstream pathways1Log in for availability1 * 1 ST1 240,00 €Specifications:MarketsourceMarketsource
- STRA219002PathDetect pFR-B-Gal Trans-reporter plasmidPathDetect pFR-B-Gal Trans-reporter plasmid50 μgb-galactosidase reporter plasmid, indicates activation of upstream pathways1Log in for availability1 * 1 ST1 110,00 €Specifications:MarketsourceMarketsource
- STRA219004PathDetect pFR-SEAP Trans-reporter PlasmdPathDetect pFR-SEAP Trans-reporter Plasmd50 μgSecreted alkaline phosphatase reporter plasmid, indicates activation of upstream pathways1Log in for availability1 * 1 ST1 110,00 €Specifications:MarketsourceMarketsource
- STRA219050PathDetect pFR-Luc plasmidPathDetect pFR-Luc plasmid50 μgLuciferase reporter plasmid, indicates activation of upstream pathways50 µGLog in for availability1 * 50 µG833,00 €Specifications:MarketsourceMarketsource
- STRA219000PathDetect c-JUN Trans-reporting systemPathDetect c-JUN Trans-reporting system50 reactionsc-Jun N-terminal Kinase System, Includes: pFR-Luc Plasmid (1 mg/ml) , pFA2-cJun Plasmid (25 ng/μl), pFC2-dbd Plasmid (25 ng/μl), and pFC-MEKK Plasmid (25 ng/μl)1Log in for availability1 * 1 ST1 480,00 €Specifications:MarketsourceMarketsource
- STRA219005PathDetect Elk1 Trans-reporting systemPathDetect Elk1 Trans-reporting system50 reactionsMAPK (mitogen-activated protein kinase) Pathway System Includes: pFR-Luc, pFA2-Elk1, pFC2-dbd and pFC-MEK1 plasmids1Log in for availability1 * 1 ST1 480,00 €Specifications:MarketsourceMarketsource
- STRA219015PathDetect CHOP Trans-reporting systemPathDetect CHOP Trans-reporting system50 reactionsp38 Kinase Pathway System, Includes: pFR-Luc Plasmid (1 mg/ml) , pFA-CHOP Plasmid (25 ng/μl), pFC2-dbd Plasmid (25 ng/μl), and pFC-MEK3 Plasmid (25 ng/μl)1Log in for availability1 * 1 ST1 480,00 €Specifications:MarketsourceMarketsource