Orders LinkContactUsLinkComponent
Cis-reporting systems, PathDetect
Cis-reporting systems, PathDetect
Marketsource
 :   STRATAGENE
Cis-reporting systems, PathDetect
Marketsource
Cis-reporting systems, PathDetect
Cis-reporting systems, PathDetect
The PathDetect cis-reporting systems is designed for rapid and convenient assessment of the in vivo activation of these pathways. Each cis-reporter plasmid contains the luciferase reporter gene driven by a basic promoter element (TATA box) joined to tandem repeats of various binding elements.
  • Identify pathway-specific transcription activation
  • Easier and faster than blotting or gel-shift assays
  • Sensitive, quantitative assays
When PathDetect cis-reporting and expression plasmids encoding the gene of interest are co-transfected into mammalian cells, increased reporter activity indicates transcription activation and involvement of your gene product in the pathway leading to the enhancer element.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
  • Description
    Details
    Contents
    Pk
  • PathDetect pFC-MEKK Control plasmid
    Positive control plasmid for c-Jun reporting system
    1
    1 * 1 ST
    1 * 1 ST - 690,00 €
    690,00 €
    1 * 1 ST
       690,00 €
    Marketsource
    Marketsource
  • PathDetect pFC-p53 plasmid
    Positive control plasmid, recommended for use with p53-Luc plasmid
    1
    1 * 1 ST
    1 * 1 ST - 690,00 €
    690,00 €
    1 * 1 ST
       690,00 €
    Marketsource
    Marketsource
  • PathDetect p53 Cis-reporting System
    System Includes: p53-Luc Cis-reporter plasmid and pFC-p53 control plasmid
    p53-Luc plasmid (1 μg/μl) and pFC-p53 positive control plasmid (25 ng/μl)
    1
    1 * 1 ST
    1 * 1 ST - 1 170,00 €
    1 170,00 €
    1 * 1 ST
       1 170,00 €
    Marketsource
    Marketsource
  • PathDetect pAP1-Luc plasmid
    PathDetect pAP-1-Luc Cis-reporter plasmid
    1
    1 * 1 ST
    1 * 1 ST - 799,00 €
    799,00 €
    1 * 1 ST
       799,00 €
    Marketsource
    Marketsource
  • PathDetect pSRE-Luc plasmid
    PathDetect pSRE-Luc Cis-reporter plasmid
    1
    1 * 1 ST
    1 * 1 ST - 799,00 €
    799,00 €
    1 * 1 ST
       799,00 €
    Marketsource
    Marketsource
  • PathDetect pSRF-Luc plasmid
    PathDetect pSRF-Luc Cis-reporter plasmid
    1
    1 * 1 ST
    1 * 1 ST - 799,00 €
    799,00 €
    1 * 1 ST
       799,00 €
    Marketsource
    Marketsource
  • PathDetect p53-Luc plasmid
    PathDetect p53-Luc Cis-reporter plasmid
    1
    1 * 1 ST
    1 * 1 ST - 799,00 €
    799,00 €
    1 * 1 ST
       799,00 €
    Marketsource
    Marketsource
  • PathDetect pISRE-Luc plasmid
    PathDetect pISRE-Luc Cis-reporter plasmid
    1
    1 * 1 ST
    1 * 1 ST - 799,00 €
    799,00 €
    1 * 1 ST
       799,00 €
    Marketsource
    Marketsource
  • PathDetect pCIS-CK plasmid
    Negative control plasmid, recommended for use with all cis-reporter plasmids
    1
    1 * 1 ST
    1 * 1 ST - 799,00 €
    799,00 €
    1 * 1 ST
       799,00 €
    Marketsource
    Marketsource
  • PathDetect pGAS-Luc plasmid
    PathDetect pGAS-Luc Cis-reporter plasmid
    1
    1 * 1 ST
    1 * 1 ST - 799,00 €
    799,00 €
    1 * 1 ST
       799,00 €
    Marketsource
    Marketsource
  • PathDetect pTARE-Luc plasmid
    The pTARE-Luc plasmid is incubated at 37 °C for 60 minutes and run on a 0.8% (w/v) agarose gel to test for the absence of contaminating nucleases.
    1
    1 * 1 ST
    1 * 1 ST - 799,00 €
    799,00 €
    1 * 1 ST
       799,00 €
    Marketsource
    Marketsource
  • PathDetect pFC-PKA Control plasmid
    PathDetect pFC-PKA plasmid
    10 µG
    1 * 10 µG
    1 * 10 µG - 690,00 €
    690,00 €
    1 * 10 µG
       690,00 €
    Marketsource
    Marketsource
  • PathDetect pLuc-MCS plasmid
    PathDetect pLuc-MCS Multiple Cloning Site Vector
    50 µG
    1 * 50 µG
    1 * 50 µG - 843,00 €
    843,00 €
    1 * 50 µG
       843,00 €
    Marketsource
    Marketsource