DNA ladders and markers
Catalog # STRA201115
Supplier: STRATAGENE
CAS Number:
Specifications
- Pk:250 µG
- Description:kB DNA ladder
- Seq:02
Specifications
About this item
The supercoiled DNA ladder contains fragments from 2 to 10 kB at 1 kB increments and an additional band at 12 kB.
- Fragments are exact multiples or fractions of 1000 bp
- Includes useful reference point for determining location
- Economical
- Easy radiolabelling of ladder
- Known mass amount of each fragment for DNA quantitation
The kB DNA ladder contains fragments from 1 to 10 kB at 1 kB increments, as well as bands at 250 bp, 500 bp, 750 bp, 1500 bp and 12 kB.
Concentration: 500 µg/ml in 10 mM Tris-HCl (pH 7,5), 1 mM EDTA.
Caution: For research use only. Not for use in diagnostic procedures.