Ethylenediamine ≥99.0% (by GC), GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:107-15-3
:
Formula:C₂H₈N₂
MDL Number:MFCD00008204
:60.1 g/mol
UN:1604
ADR:8,II
:GHS02
:GHS06
:GHS05
:GHS08
Specification Test Results
- Assay (GC):≥ 99.0%
- Appearance:Clear liquid
- Colouration:≤ 15 APHA
- Water :≤ 0.5%
Specification Test Results
Related Information
Certificates
Search by Lot Number35591387.pdf spec_VWRC88382.180 spec_VWRC88382.290 spec_VWRC88382.320 spec_VWRC88382.410
Product specification
More