Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trifluoroacetic acid
Trifluoroacetic acid
Trifluoroacetic acid
Trifluoroacetic acid
Catalog # 80457-10ML
Supplier:
Merck
CAS Number:
Expand image
Trifluoroacetic acid
Catalog # 80457-10ML
Supplier:
Merck
CAS Number:
Specifications
Pk:
10 mL
Seq:
2275
Specifications
About this item
Product Details
:
:
114.02 g/mol
CAS Number:
76-05-1
Formula:
C₂HF₃O₂
MDL Number:
MFCD00004169
Product Details