
Celite® 545, filter aid, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:68855-54-9
:
Formula:SiO₂
MDL Number:MFCD00132803
:60.08 g/mol
:GHS07
Specification Test Results
- Identification:Passes test
- pH (25°C; 10%):≥ 8,5
- Loss on ignition (600 °C):≤ 0.6%
- Particle size (< 0.1 mm):≥ 60%
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC22557.230 spec_VWRC22557.290 spec_VWRC22557.360 spec_VWRC22557.460
Product specification
VWR® for Drying and Filtration