Order Entry
Germany
ContactUsLinkComponent
J.T.Baker® BAKER INSTRA-ANALYZED®, Titanium Plasma Standards
J.T.Baker® BAKER INSTRA-ANALYZED®, Titanium Plasma Standards
  5752.0100
 :  Avantor
 :  
J.T.Baker® BAKER INSTRA-ANALYZED®, Titanium Plasma Standards
  5752.0100
 :  Avantor
 :  

 

  • Concentration:
    10000 mg/L
  • Matrix:
    2-5% HNO₃+tr. HF
  • Pk:
    100 mL
  • Seq:
    2
  • Element:
    Titanium
  • Symbol:
    Ti