Order Entry
Germany
ContactUsLinkComponent
Yeast recombinant thioreduxin reductase (NAPH)
Yeast recombinant thioreduxin reductase (NAPH)
Supplier:  Apollo Scientific
Certificates
Yeast recombinant thioreduxin reductase (NAPH)
Supplier:  Apollo Scientific

Specifications

  • Enzyme type:
    Recombinant
  • Species:
    Yeast
  • Storage conditions:
    Ambient
  • Enzyme name:
    Thioreduxin Reductase (NAPH)
  • Enzyme synonyms:
    TrxR

Specifications

Frequently Bought Together