L-Asparaginase
Supplier: Apollo Scientific
About this item
3 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- APOSBITP1331-10KIU
- APOSBITP1331-500IU
- APOSBITP1331-2500I
- Enzyme name:
- L-Asparaginase
- L-Asparaginase
- MDL No.:
- MFCD00130565
- MFCD00130565
- CAS No.:
- 9015-68-3
- 9015-68-3
- Storage conditions:
- Refrigerator
- Refrigerator
Specifications
Product Family Options
Product Information
- Pk InfoPkAvailabilityPrice
- Specifications:New Product
More
Specifications
Cat. No.APOSBITP1331-10KIUEnzyme nameMDL No.CAS No.Storage conditionsNew Product - Specifications:
More
Specifications
Cat. No.APOSBITP1331-500IUEnzyme nameL-AsparaginaseMDL No.MFCD00130565CAS No.9015-68-3Storage conditionsRefrigerator - Specifications:
More
Specifications
Cat. No.APOSBITP1331-2500IEnzyme nameL-AsparaginaseMDL No.MFCD00130565CAS No.9015-68-3Storage conditionsRefrigerator
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



