Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Horseradish peroxidase
Horseradish peroxidase
:
Apollo Scientific
Horseradish peroxidase
:
Apollo Scientific
Species:
Horseradish
Storage conditions:
Refrigerator
MDL No.:
MFCD00071339
Enzyme name:
Peroxidase
Enzyme synonyms:
HRP