Recombinant disulfide oxidoreductase
Catalog # APOSBITP1318-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Enzyme type:Recombinant
- Storage conditions:Ambient
- MDL No.:MFCD00163364
- Enzyme name:Disulfide Oxidoreductase
- Enzyme synonyms:DsbA