Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Pepsin
Pepsin
Pepsin
Supplier:
Apollo Scientific
Certificates
Expand image
Pepsin
Supplier:
Apollo Scientific
Certificates
About this item
Specifications
Storage conditions:
Refrigerator
MDL No.:
MFCD00081840
Enzyme name:
Pepsin
Specifications
Related Information
Documentation
Certificates
Search by Lot Number
Documentation