Order Entry
Germany
ContactUsLinkComponent
Aspergillus niger Cellulase
Aspergillus niger Cellulase
Supplier:  Apollo Scientific
Certificates
Aspergillus niger Cellulase
Supplier:  Apollo Scientific
Certificates

Specifications

  • Species:
    Aspergillus niger
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00081510
  • Enzyme name:
    Cellulase

Specifications

Related Information