Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Sheep Recombinant Leptin tA
Sheep Recombinant Leptin tA
Sheep Recombinant Leptin tA
Catalog # APOSBITP1209-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Sheep Recombinant Leptin tA
Catalog # APOSBITP1209-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Sheep
Protein synonyms:
Leptin Triple Antagonist
Protein/peptide name:
Leptin tA
Specifications
About this item
Product Details
Product Details