Orders LinkContactUsLinkComponent
Multiports, b.safe
Supplier:   Bohlender
Multiports, b.safe
Supplier:  Bohlender
Certificates
Multiports, b.safe
Multiports, b.safe
8 Options Available Below
Product Details & Documents
Extension of the number of capillary and/or tubing connectors on a b.safe Waste Cap. With PFA Fittings for capillaries with Ext.Ø 1,6/2,2/3,2 mm (thread UNF ¹/₄"), 1 hose connector for tubing Int.Ø 5,5 to 8,0 mm (thread NPT ¹/₈").
  • Excellent chemical resistance
  • Available for a wide range of hose diameters
  • The conical shape gets the connection tight and leak-free
Adaptor to enlarge the number of capillary and tubing connectors of a b.safe waste cap.
Product Family Options
Product Information
  • With
    Thread
    Availability
    Price
  • 590-0183
    Tubing connector
    Tubing connector
    2× capillary connector, 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm
    NPT ¹/₈"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,20 €
    36,20 €
    1 * 1 ST
       36,20 €
  • 590-0188
    Tubing connector
    Tubing connector
    2× capillary connector, 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm, 1 hose connector for tubing Int.Ø 5,5 - 8,0 mm
    NPT ¹/₈"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57,80 €
    57,80 €
    1 * 1 ST
       57,80 €
  • 590-0184
    Tubing connector
    Tubing connector
    3× capillary connector, 3× fittings for tubing Ext.Ø 1,6 mm, 3× fittings for tubing Ext.Ø 2,2 mm, 3× fittings for tubing Ext.Ø 3,2 mm
    NPT ¹/₈"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,40 €
    36,40 €
    1 * 1 ST
       36,40 €
  • 590-0187
    Tubing connector
    Tubing connector
    3× capillary connector, 3× fittings for tubing Ext.Ø 1,6 mm, 3× fittings for tubing Ext.Ø 2,2 mm, 3× fittings for tubing Ext.Ø 3,2 mm
    NPT ¹/₈"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57,90 €
    57,90 €
    1 * 1 ST
       57,90 €
  • BOHLM509-34
    Tubing connector
    Tubing connector
    3× hose connectors for tubing Int.Ø 5,5 - 8,0 mm
    NPT ¹/₈"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,80 €
    49,80 €
    1 * 1 ST
       49,80 €
  • BOHLM509-36
    Tubing connector
    Tubing connector
    3× hose connectors for tubing Int.Ø 5,5 - 8,0 mm
    NPT ¹/₈"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57,20 €
    57,20 €
    1 * 1 ST
       57,20 €
  • 590-0186
    Tubing connector
    Tubing connector
    7× capillary connector, 7× fittings for tubing Ext.Ø 1,6 mm, 7× fittings for tubing Ext.Ø 2,2 mm, 7× fittings for tubing Ext.Ø 3,2 mm, 1 hose connector for tubing Int.Ø 5,5 - 8,0 mm
    NPT ¹/₈"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 112,00 €
    112,00 €
    1 * 1 ST
       112,00 €
  • 590-0185
    Tubing connector
    Tubing connector
    8× capillary connector, 8× fittings for tubing Ext.Ø 1,6 mm, 8× fittings for tubing Ext.Ø 2,2 mm, 8× fittings for tubing Ext.Ø 3,2 mm
    NPT ¹/₈"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 113,00 €
    113,00 €
    1 * 1 ST
       113,00 €