Order Entry
Germany
Orders LinkContactUsLinkComponent
Suction filter, b.safe

Suction filter, b.safe

Suction filter, 20 µm, 3,2 mm (¹/₈") mm
Catalog # 590-0201
Supplier:   Bohlender
Suction filter, b.safe
Catalog # 590-0201
Supplier:  Bohlender
Suction filter, b.safe
Suction filter, b.safe

Suction filter, 20 µm, 3,2 mm (¹/₈") mm

Price
68,30 €
Log in for availability
1 * 10 ST
Product Details & Documents

Filter candle made of porous PE.

  • With a bore to be put on capillary tubing
  • Restrains pollution from solvents
  • For constant analysis results

Exchange the filter regularly every three months for reliable operation.

Specifications
  • Pore size:
    20 µm
  • Description:
    Suction filter
  • For tube O-Ø:
    3,2 mm (¹/₈") mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.