Order Entry
Germany
ContactUsLinkComponent
VWR®, Red Taq DNA polymerase
VWR®, Red Taq DNA polymerase
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Red Taq DNA polymerase
Supplier:  VWR Chemicals
Certificates

Specifications

  • Application:
    Standard amplification
  • Format:
    Enzyme with/without buffer
  • High fidelity:
    No
  • Hot start:
    No
  • Storage conditions:
    – 20 °C (long-term), + 4 °C (6 months)
  • Amplicon size:
    ≤5 kb
  • Processivity:
    Standard
  • Exonuclease activity:
    5’→3’
  • Product overhang:
    3'A
  • Features:
    Visible dye
  • Buffer composition:
    10X Key Buffer, 10X Extra Buffer and 25 mM MgCl₂
  • Concentration:
    5 U/µl
  • Description:
    Taq RED DNA Polymerase 5 U/µl 1.000 units
  • Size:
    2×0,1 ml + 6×1,5 ml
  • Units:
    1000

Specifications