Capillary cutter, b.safe
Ideal for cutting capillaries and elastic tubes without reinforcement.
- Capillary cutter for up to tubing Ext.Ø 8 mm with guide bores
- High qualitiy capillary cutter
- Replacement blades available
The guide bores help to get a clean cut of the capillaries.
Ideal for cutting capillaries and elastic tubes without reinforcement.
- Capillary cutter for up to tubing Ext.Ø 8 mm with guide bores
- High qualitiy capillary cutter
- Replacement blades available
The guide bores help to get a clean cut of the capillaries.
Product Family Options
- Description
- Capillary cutter with guide bores for tubing up to Ext.Ø 8 mm1 * 1 ST53,60 €
- Replacement blades for capillary cutter for tubing up to Ext.Ø 8 mm1 * 1 ST9,20 €