Orders LinkContactUsLinkComponent
Spouts, b.safe
Spouts, b.safe
Supplier:   Bohlender
Spouts, b.safe
Supplier:  Bohlender
Certificates
Spouts, b.safe
7 Options Available Below
Product Details & Documents
Screw cap for canister thread, freely turnable spout tube, rigid or flexible design with integrated safety aeration for constant flow.
  • Spout tube is freely turnable
  • Integrated safety aeration
  • Prevents loss of fluids
Quick refilling of liquids without dripping directly from canister.
Product Family Options
Product Information
  • Thread
    Material
    Availability
    Price
  • 590-0322
    Spout tube flexible
    Spout tube flexible
    GL 45
    PE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,00 €
    17,00 €
    1 * 1 ST
       17,00 €
  • 590-0323
    Spout tube flexible
    Spout tube flexible
    S55
    PE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,90 €
    17,90 €
    1 * 1 ST
       17,90 €
  • 590-0324
    Spout tube flexible
    Spout tube flexible
    S60/61
    PE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,20 €
    19,20 €
    1 * 1 ST
       19,20 €
  • 590-0325
    Spout tube flexible
    Spout tube flexible
    S70/71
    PE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,00 €
    15,00 €
    1 * 1 ST
       15,00 €
  • 590-0326
    Spout tube rigid
    Spout tube rigid
    S55
    PE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,00 €
    15,00 €
    1 * 1 ST
       15,00 €
  • 590-0327
    Spout tube rigid
    Spout tube rigid
    S60/61
    PE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,00 €
    17,00 €
    1 * 1 ST
       17,00 €
  • 590-0328
    Spout tube rigid, static dissipative
    Spout tube rigid, static dissipative
    S60/61
    PE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 147,00 €
    147,00 €
    1 * 1 ST
       147,00 €