


2,4,5-Trifluorophenol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2268-16-8
:
Formula:C₆H₃F₃O
MDL Number:MFCD00061215
:148.08 g/mol
Melting Pt:37…42 °C
Flash Pt:55 °C
UN:2925
ADR:4.1+8,III
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient