0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
4-Bromo-2-fluoro-6-nitroanisole
4-Bromo-2-fluoro-6-nitroanisole
4-Bromo-2-fluoro-6-nitroanisole
:
Apollo Scientific
4-Bromo-2-fluoro-6-nitroanisole
:
Apollo Scientific
:
74266-66-3
:
:
C₇H₅BrFNO₃
:
MFCD00013374
:
250.02 g/mol
:
53…56 °C
:
GHS07
:
Ambient