2-({2-Oxo-2-[4-(trifluoromethyl)anilino]ethyl}sulphanyl)acetic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:338421-16-2
:
Formula:C₁₁H₁₀F₃NO₃S
MDL Number:MFCD00202307
:293.27 g/mol
Melting Pt:121…123 °C
Storage Temperature:Ambient