Methyl-2-{[4-formyl-1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]sulphanyl}benzenecarboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:321553-50-8
:
Formula:C₁₄H₁₁F₃N₂O₃S
MDL Number:MFCD00140853
:344.31 g/mol
Melting Pt:158…160 °C
Storage Temperature:Ambient