2-Bromo-α,α,α,4-tetrafluorotoluene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:351003-21-9
:
Formula:C₇H₃BrF₄
MDL Number:MFCD03094041
:243 g/mol
Boiling Pt:161…162 °C
Density:1.753 g/cm³ (25 °C)
Flash Pt:78.3 °C
:GHS07
Storage Temperature:Ambient