Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(4-Fluorobenzenesulphonyl)phenylalanine
N-(4-Fluorobenzenesulphonyl)phenylalanine
N-(4-Fluorobenzenesulphonyl)phenylalanine
N-(4-Fluorobenzenesulphonyl)phenylalanine
Supplier:
Apollo Scientific
Certificates
Expand image
N-(4-Fluorobenzenesulphonyl)phenylalanine
Supplier:
Apollo Scientific
About this item
CAS Number:
40279-96-7
:
Formula:
C₁₅H₁₄FNO₄S
MDL Number:
MFCD00173011
:
323.35 g/mol
Melting Pt:
108…110 °C
Storage Temperature:
Ambient