Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(4-Fluorophenyl)biguanide hydrochloride
1-(4-Fluorophenyl)biguanide hydrochloride 97%
1-(4-Fluorophenyl)biguanide hydrochloride 97%
1-(4-Fluorophenyl)biguanide hydrochloride 97%
Supplier:
Apollo Scientific
Certificates
Expand image
1-(4-Fluorophenyl)biguanide hydrochloride 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
16018-83-0
:
Formula:
C₈H₁₁ClFN₅
MDL Number:
MFCD00051986
:
231.66 g/mol
Melting Pt:
212…216 °C
:
GHS07
Storage Temperature:
Ambient