Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4,5-Trifluoro-3-methylbenzoic acid
2,4,5-Trifluoro-3-methylbenzoic acid 95%
2,4,5-Trifluoro-3-methylbenzoic acid 95%
2,4,5-Trifluoro-3-methylbenzoic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,4,5-Trifluoro-3-methylbenzoic acid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
112822-85-2
:
Formula:
C₈H₅F₃O₂
MDL Number:
MFCD04972492
:
190.1193 g/mol
Storage Temperature:
Ambient